Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUBGCP2 Antibody (GCP2-01), HRP, Novus Biologicals™
SDP

Catalog No. NB355724 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355724 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355724 Supplier Novus Biologicals Supplier No. NBP221793H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TUBGCP2 Monoclonal antibody specifically detects TUBGCP2 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen TUBGCP2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GCP2-01
Conjugate HRP
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias Gamma-ring complex protein 103 kDa, gamma-tubulin complex component 2, gamma-tubulin complex protein 2, GCP-2, GCP2hGrip103, h103p, hGCP2, hSpc97, MGC138162, SPBC97, Spc97p, Spindle pole body protein Spc97 homolog, tubulin, gamma complex associated protein 2
Host Species Mouse
Immunogen GST-fusion protein containing amino acids 2-194 of mouse GCP2.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10844
Target Species Human, Mouse, Rat, Pig
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.