Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Tubular Spring Balances PROMO
Shop All Eisco Products

Click to view available options
Capacity:
100 g/1 N
Description
- Made from plastic
- Color coded for convenience
- Spring mechanism clearly visible
- Zero adjustment incorporated
Specifications
Specifications
Color | Black |
Grade | 9 to 12 |
Material | Plastic |
Capacity | 100 g/1 N |
Languages | English |
Display Type | Analog |
Type | Tubular Spring Balances |
Legal for Trade | No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction