Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tubulin alpha-1B Antibody (OTI3G3), FITC, Novus Biologicals™
SDP

Catalog No. NBP274703F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274703F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274703F Supplier Novus Biologicals Supplier No. NBP274703F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Tubulin alpha-1B Monoclonal specifically detects Tubulin alpha-1B in Human, Mouse samples. It is validated for Immunohistochemistry.

Specifications

Antigen Tubulin alpha-1B
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI3G3
Conjugate FITC
Dilution Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias alpha tubulin, Alpha-tubulin ubiquitous, K-ALPHA-1, tubulin alpha-1B chain, Tubulin alpha-ubiquitous chain, Tubulin K-alpha-1, tubulin, alpha 1b, tubulin, alpha, ubiquitous
Gene Symbols TUBA1B
Host Species Mouse
Immunogen Full length recombinant protein of human Tubulin alpha-1B (NP_006073) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Biology, Cytoskeleton Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10376.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.