Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tubulin alpha-3E Antibody (OTI2A8), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N274704P594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274704P594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274704P594 Supplier Novus Biologicals Supplier No. NBP274704PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Tubulin alpha-3E Monoclonal specifically detects Tubulin alpha-3E in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen Tubulin alpha-3E
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2A8
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Alpha-tubulin 3E, tubulin alpha-3E chain, tubulin, alpha 3e
Gene Symbols TUBA3E
Host Species Mouse
Immunogen Full length recombinant protein of human Tubulin alpha-3E (NP_997195) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 112714.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.