Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tubulin alpha-8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP174176
Description
Tubulin alpha-8 Polyclonal specifically detects Tubulin alpha-8 in Mouse samples. It is validated for Western Blot.Specifications
Tubulin alpha-8 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
Alpha-tubulin 8, TUBAL2Tubulin alpha chain-like 2, tubulin alpha-8 chain, tubulin, alpha 8 | |
Rabbit | |
50 kDa | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: ; Dog: 86%; Pig: 86%; Guinea pig: 86%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q9JJZ2 | |
TUBA8 | |
Synthetic peptides corresponding to the N terminal of Tuba8. Immunizing peptide sequence ADGTFGTQASKINDDDSFTTFFSETGNGKHVPRAVMVDLEPTVVDEVRAG. | |
Affinity purified | |
RUO | |
51807 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction