Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Tubulin Tracker™ Deep Red

Click to view available options
Quantity:
300 slides
60 slides
Description
- Based on Docetaxel conjugated with a bright, photostable deep-red fluorophore
- Docetaxel belongs to the family of cytoskeletal drugs that target tubulin
- Absorbs and emits optimally at 652 nm and 669 nm, respectively and can be visualized with standard Cy5 filter settings using almost any fluorescent imaging instrument
- Can be multiplexed with blue, green, orange, red, and near-IR fluorophores.

Specifications
Specifications
Content And Storage | Contains sufficient material for at least 60 slides at 1 mL per slide. •1 vial Tubulin Tracker Deep Red (Add 60 μL/vial of anhydrous DMSO to make a 1000X (1mM) stock solution. Dissolve stock solution 1:1000 in cell imaging solution to make 1X (1 μM) staining solution.) •1 x 77 mg Probenecid (Dissolve vial content in 1 mL of live cell compatible buffer to make a 100X stock solution.) Store at ≤–20°C after recieving. Desiccate and protect from light. When stored as directed, product is stable for at least 6 months. |
Excitation Wavelength Range | 652⁄669 |
Quantity | 60 slides |
Product Line | TubulinTracker™ |
Shipping Condition | Approved for shipment on Wet or Dry Ice |
Label Type | UV-Vis Labels |
Product Type | Tubulin Tracker |
SubCellular Localization | Tubulin |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction