Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynalon™ Tuff Vials
SCI_ED

Catalog No. p-6966014 Shop All Dynalab Corporation Products
Click to view available options
Volume (Metric):
5 mL
10 mL

Catalog No. S37270

Supplier: Dynalon™ 6264340005

Only null left
Add to Cart
Add to Cart

Ideal for leakproof shipping of reagents and buffer solutions, for packaging of serum, veterinary, pharmaceuticals, cosmeceutical and nutraceutical extracts. Dynalon™ Tuff Vials are used with most aromatic compounds and essential oils.

  • Vials are made of break resistant polycarbonate
  • Includes polypropylene leakproof closures
  • Reusable and autoclavable
  • Clear glass helps viewing of valuable contents, liquid or solid
  • 180mL can also be used as an outer protective container

Specifications

Material Polycarbonate
Volume (Metric) 5 mL
Style Round
Closure Material Polypropylene
Closure Size 20-415
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.