Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUG Antibody (4A11A6G11) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP147360 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP147360 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP147360 Supplier Novus Biologicals Supplier No. NBP147360
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TUG Monoclonal specifically detects TUG in Mouse samples. It is validated for Western Blot, ELISA.

Specifications

Antigen TUG
Applications Western Blot, ELISA
Classification Monoclonal
Clone 4A11A6G11
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1: 500 - 1:2000, ELISA 1:10000
Gene Alias Alveolar soft part sarcoma chromosomal region candidate gene 1 protein, alveolar soft part sarcoma chromosome region, candidate 1, Alveolar soft part sarcoma locus, ASPLUBX domain-containing protein 9, ASPSrenal cell carcinoma gene on chromosome 17, RCC17renal cell carcinoma, papillary, 17, Renal papillary cell carcinoma protein 17, tether containing UBX domain for GLUT4, TUG, UBX domain protein 9, UBXD9ASPCR1, UBXN9FLJ45380
Gene Symbols ASPSCR1
Host Species Mouse
Immunogen Purified recombinant fragment of TUG expressed in E. Coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79058
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.