Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TULA/STS-2 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB064843 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB064843 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB064843 Supplier Novus Biologicals Supplier No. NBP300200G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TULA/STS-2 Polyclonal specifically detects TULA/STS-2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen TULA/STS-2
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Cbl-interacting protein 4, gene similar to UBA containing SH3 domain10CLIP4STS-2T-cell ubiquitin ligand protein, STS2, Suppressor of T-cell receptor signaling 2, T-cell ubiquitin ligand, TULAubiquitin-associated and SH3 domain-containing protein A, ubiquitin associated and SH3 domain containing A
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant TULA/STS-2 (AAH69511.1; Ala354-Asn623).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53347
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.