Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Tunicamycin
GSA_VAGREENER_CHOICESDP

Catalog No. 351610 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-161-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 35-161-0 Supplier R&D Systems™ Supplier No. 3516/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Antibiotic; GlcNAc phosphotransferase inhibitor

Tunicamycin is an antibiotic; inhibits GlcNAc phosphotransferase (GPT). Blocks the formation of N-glycosidic linkages by inhibiting the first step in glycoprotein synthesis. Activity induces ER stress and causes G1 arrest; can be used to induce autophagy. Tunicamycin contains four main components as follows: Homolog A, n=8, C37H60N4O16, molecular weight = 816.90 Homolog B, n=9, C38H62N4O16, molecular weight = 830.93 Homolog C, n=10, C39H64N4O16, molecular weight = 844.95 Homolog D, n=11, C40H66N4O16, molecular weight = 858.99 The composition of this product will vary from batch to batch and can be found on the relevant certificate of analysis.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 11089-65-9

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 844.95
Product Type Tunicamycin
Purity >98%
Chemical Name or Material Tunicamycin
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.