Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TUT1 Antibody, Novus Biologicals™
SDP

Catalog No. NB435344 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB435344 25 μL
NBP257861 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB435344 Supplier Novus Biologicals Supplier No. NBP25786125UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TUT1 Polyclonal specifically detects TUT1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TUT1
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1-4 μg/mL
Formulation PBS (pH 7.2), 40% Glycerol with 0.02% Sodium Azide
Gene Alias EC 2.7.7.19, EC 2.7.7.52, FLJ21850, FLJ22267, FLJ22347, MGC131987, MGC149809, nuclear speckle targeted phosphatidylinositol 4-phosphate 5-kinase type I-alpharegulated-poly(A) polymerase, nuclear speckle-targeted PIPK1A-regulated-poly(A) polymerase, PAP-associated domain-containing 2, PAPD2, poly(A) polymerase associated domain containing 2, RBM21, RNA binding motif protein 21, RNA uridylyltransferase, RNA-binding motif protein 21, RNA-binding protein 21, speckle targeted PIP5K1A-regulated poly(A) polymerase, star-PAP, STARPAP, terminal uridylyl transferase 1, U6 snRNA-specific, TUTase, U6 snRNA-specific terminal uridylyltransferase 1, U6 TUTase, U6-TUTase
Gene Symbols TUT1
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LASPQSLPPASPLLEDREEGDLGKASELAETPKEEKAEGAAMLELVGSILRGCVPGVYRVQTVPSARRPVVKFCHRPSGLHGD
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64852
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.