Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TWEAK R/TNFRSF12 Antibody (7H8), Novus Biologicals™
SDP

Catalog No. NB298417 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298417 100 μL
NB298416 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298417 Supplier Novus Biologicals Supplier No. NBP326280100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

TWEAK R/TNFRSF12 Monoclonal antibody specifically detects TWEAK R/TNFRSF12 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen TWEAK R/TNFRSF12
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 7H8
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias CD266, CD266 antigen, FGF-inducible 14, Fibroblast growth factor-inducible immediate-early response protein 14, FN14tumor necrosis factor receptor superfamily member 12A, tumor necrosis factor receptor superfamily, member 12A, TweakR, Tweak-receptor, type I transmembrane protein Fn14
Host Species Rabbit
Immunogen A synthesized peptide derived from Human TWEAK R/TNFRSF12 [UniProt Q9NP84]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Biologically Active Proteins, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 51330
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.