Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TWEAK R/TNFRSF12 Antibody (ITEM-4), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP200409FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP200409FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP200409FR Supplier Novus Biologicals Supplier No. NBP200409FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TWEAK R/TNFRSF12 Monoclonal specifically detects TWEAK R/TNFRSF12 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen TWEAK R/TNFRSF12
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone ITEM-4
Conjugate DyLight 680
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Gene Alias CD266, CD266 antigen, FGF-inducible 14, Fibroblast growth factor-inducible immediate-early response protein 14, FN14tumor necrosis factor receptor superfamily member 12A, tumor necrosis factor receptor superfamily, member 12A, TweakR, Tweak-receptor, type I transmembrane protein Fn14
Gene Symbols TNFRSF12A
Host Species Mouse
Immunogen The immunogen for this antibody is TWEAKR.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 51330
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.