Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TWEAK/TNFSF12 Antibody (MM0590-10T24), FITC, Novus Biologicals™
SDP

Catalog No. NB344320 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344320 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344320 Supplier Novus Biologicals Supplier No. NBP211960F

Mouse Monoclonal Antibody

TWEAK/TNFSF12 Monoclonal antibody specifically detects TWEAK/TNFSF12 in Human samples. It is validated for Western Blot

Specifications

Antigen TWEAK/TNFSF12
Applications Western Blot
Classification Monoclonal
Clone MM0590-10T24
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias APO3 ligand, APO3LMGC20669, DR3LGTWEAKAPO3/DR3 ligand, MGC129581, TNF-related weak inducer of apoptosis, tumor necrosis factor (ligand) superfamily, member 12, tumor necrosis factor ligand superfamily member 12
Host Species Mouse
Immunogen Human recombinant TWEAK/TNFSF12
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8742
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.