Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Twist-1 Antibody (927403R), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB376831 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB376831 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB376831 Supplier Novus Biologicals Supplier No. FAB6230RI
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Twist-1 Monoclonal specifically detects Twist-1 in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Western Blot.

Specifications

Antigen Twist-1
Applications CyTOF, Flow Cytometry (Intracellular Staining), Western Blot
Classification Monoclonal
Clone 927403R
Conjugate Janelia Fluor 549
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias acrocephalosyndactyly 3, ACS3, B-HLH DNA binding protein, BHLHA38, bHLHa38BPES3, Class A basic helix-loop-helix protein 38, CRS1, H-twistblepharophimosis, epicanthus inversus and ptosis 3, SCSBPES2, TWIST, twist homolog 1 (Drosophila), TWIST homolog of drosophila, twist-related protein 1
Host Species Mouse
Immunogen E.coli-derived recombinant human Twist-1, Met1-His202, Accession # Q15672
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7291
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.