Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Twist-2 Antibody (OTI6H10), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP274724G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274724G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274724G Supplier Novus Biologicals Supplier No. NBP274724G

Mouse Monoclonal Antibody

Twist-2 Monoclonal specifically detects Twist-2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Twist-2
Applications Western Blot
Classification Monoclonal
Clone OTI6H10
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BHLHA39, bHLHa39twist-related bHLH protein Dermo1, Class A basic helix-loop-helix protein 39, Dermis-expressed protein 1, Dermo-1, DERMO1MGC117334, twist homolog 2 (Drosophila), twist-related protein 2
Gene Symbols TWIST2
Host Species Mouse
Immunogen Full length recombinant protein of human Twist-2 (NP_476527) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Inhibitors Activators and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 117581.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.