Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TWISTNB Antibody, Novus Biologicals™
SDP

Catalog No. NBP227081 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227081 0.1 mg
NBP227081SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227081 Supplier Novus Biologicals Supplier No. NBP227081
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TWISTNB Polyclonal specifically detects TWISTNB in Human, Mouse, Equine, Primate samples. It is validated for Western Blot.

Specifications

Antigen TWISTNB
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 3-5ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q3B726
Gene Alias DNA-directed RNA polymerase I subunit RPA43, TWIST neighbor, Twist neighbor protein
Gene Symbols TWISTNB
Host Species Rabbit
Immunogen A portion of amino acids 50-128 of human Twistnb was used as the immunogen. 61 78
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221830
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, horse and chimpanzee, and 94% homologous in mouse, rat and dog.
Target Species Human, Mouse, Equine, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.