Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Two pore calcium channel protein 2 Antibody - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018496 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018496 0.1 mg
NBP130670 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018496 Supplier Novus Biologicals Supplier No. NBP1306700.1MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Two pore calcium channel protein 2 Polyclonal specifically detects Two pore calcium channel protein 2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Two pore calcium channel protein 2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:200-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:200-1:2000
Formulation Sterilized water with No Preservative
Gene Accession No. Q8BWC0
Gene Alias FLJ41094, SHEP10, TPC2two pore calcium channel protein 2, two pore segment channel 2, two-pore calcium channel protein 2, Voltage-dependent calcium channel protein TPC2
Gene Symbols TPCN2
Host Species Rabbit
Immunogen A synthetic peptide from an extracellular domain of human TPC2 (Two pore calcium channel protein 2) conjugated to an immunogenic carrier protein was used as the immunogen.
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219931
Test Specificity Human. Other species not yet tested.
Reconstitution Reconstitute with 0.5 ml sterilized water.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.