Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Two pore calcium channel protein 2 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | Two pore calcium channel protein 2 |
---|---|
Concentration | LYOPH |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Two pore calcium channel protein 2 Polyclonal specifically detects Two pore calcium channel protein 2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Two pore calcium channel protein 2 | |
Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q8BWC0 | |
219931 | |
IgG | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
LYOPH | |
Polyclonal | |
Purified | |
RUO | |
Sterilized water with No Preservative | |
FLJ41094, SHEP10, TPC2two pore calcium channel protein 2, two pore segment channel 2, two-pore calcium channel protein 2, Voltage-dependent calcium channel protein TPC2 | |
TPCN2 | |
Primary | |
Human. Other species not yet tested. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title