Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Two pore calcium channel protein 2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP145234 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP145234 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP145234 Supplier Novus Biologicals Supplier No. NBP145234
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Two pore calcium channel protein 2 Polyclonal specifically detects Two pore calcium channel protein 2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen Two pore calcium channel protein 2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/ml, Immunohistochemistry 3.75 ug/ml, Immunohistochemistry-Paraffin 3.75 ug/ml, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_620714.2
Gene Alias FLJ41094, SHEP10, TPC2two pore calcium channel protein 2, two pore segment channel 2, two-pore calcium channel protein 2, Voltage-dependent calcium channel protein TPC2
Gene Symbols TPCN2
Host Species Goat
Immunogen Peptide with sequence C-EDELTERLSQHPH corresponding to C-Terminus according to NP_620714.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219931
Test Specificity This antibody is specific to the C-Terminus.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.