Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TXLNA Polyclonal Antibody
Rabbit Polyclonal Antibody
$178.40 - $511.50
Specifications
Antigen | TXLNA |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119453
|
Invitrogen™
PA5119453 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121514
|
Invitrogen™
PA5121514 |
20 μL |
Each for $178.40
|
|
|||||
Description
TXLNA Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
May be involved in intracellular vesicle traffic and potentially in calcium-dependent exocytosis in neuroendocrine cells. [UniProt].Specifications
TXLNA | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
TXLNA | |
Alpha-taxilin; DKFZp451J0118; il 14; IL14; IL-14; interleukin 14; MGC118870; MGC118871; RP4-622L5.4; taxilin alpha; TXLN; TXLNA | |
TXLNA | |
Primary | |
Antigen affinity chromatography, Protein A |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
P40222 | |
200081 | |
Recombinant protein Taxilin Alpha. The antigen corresponds to amino acid range 328-531 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title