Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TXNDC Antibody (OTI3H5), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP27472A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27472A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27472A30 Supplier Novus Biologicals Supplier No. NBP274725AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TXNDC Monoclonal specifically detects TXNDC in Human, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen TXNDC
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI3H5
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias DKFZp564E1962, PDIA11, protein disulfide isomerase family A, member 11, thioredoxin domain containing 1, thioredoxin domain-containing, Thioredoxin domain-containing protein 1, thioredoxin-related transmembrane protein 1, TMXTXNDC, Transmembrane Trx-related protein, TXNDC1thioredoxin domain containing
Gene Symbols TMX1
Host Species Mouse
Immunogen Full length recombinant protein of human TXNDC (NP_110382) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 81542
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.