Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TXNDC5 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB098194 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB098194 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB098194 Supplier Novus Biologicals Supplier No. NBP298558AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TXNDC5 Polyclonal specifically detects TXNDC5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen TXNDC5
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50 mM Sodium Borate
Gene Alias ENDOPDI, ERP46, FLJ90810, HCC-2, member 15, MGC3178, PDIA15, thioredoxin domain containing 5, thioredoxin domain containing 5 (endoplasmic reticulum), thioredoxin domain-containing protein 5, thioredoxin related protein, TLP46
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human TXNDC5 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 81567
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.