Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TXNDC5 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001452 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001452 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001452 Supplier Novus Biologicals Supplier No. NB1001452
Add to Cart
Add to Cart

Goat Polyclonal Antibody

TXNDC5 Polyclonal specifically detects TXNDC5 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot.

Specifications

Antigen TXNDC5
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.01 - 0.1 ug/mL, Flow Cytometry 10 ug/mL, Immunocytochemistry/Immunofluorescence 10 ug/mL, Immunohistochemistry-Paraffin 3.75 ug/mL, Peptide ELISA Detection limit 1:64000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_110437
Gene Alias ENDOPDI, ERP46, FLJ90810, HCC-2, member 15, MGC3178, PDIA15, thioredoxin domain containing 5, thioredoxin domain containing 5 (endoplasmic reticulum), thioredoxin domain-containing protein 5, thioredoxin related protein, TLP46
Gene Symbols TXNDC5
Host Species Goat
Immunogen Peptide with sequence C-SLHRFVLSQAKDEL corresponding to C-Terminus according to NP_110437.2, NP_001139021.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 81567
Test Specificity This antibody is expected to be able to recognize both reported human isoforms, as represented by NP_110437.2; NP_001139021.1.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.