Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TXNDC5 Antibody (OTI2E6), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP274726A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274726A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274726A30 Supplier Novus Biologicals Supplier No. NBP274726AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TXNDC5 Monoclonal specifically detects TXNDC5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen TXNDC5
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2E6
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias ENDOPDI, ERP46, FLJ90810, HCC-2, member 15, MGC3178, PDIA15, thioredoxin domain containing 5, thioredoxin domain containing 5 (endoplasmic reticulum), thioredoxin domain-containing protein 5, thioredoxin related protein, TLP46
Gene Symbols TXNDC5
Host Species Mouse
Immunogen Full length recombinant protein of human TXNDC5 (NP_071368) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 81567.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.