Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TXNIP Antibody (JY2) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB014074 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB014074 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB014074 Supplier Novus Biologicals Supplier No. NBP281001
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TXNIP Monoclonal specifically detects TXNIP in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Proximity Ligation Assay, Immunocytochemistry, CyTOF-ready, Knockout Validated.

Specifications

Antigen TXNIP
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In situ PLA, Immunocytochemistry, CyTOF, Immunoassay
Classification Monoclonal
Clone JY2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot : 1:1000, Flow Cytometry : 2-5 ug/0.1x10^6 cells, Immunohistochemistry, Immunoprecipitation : 5x10^6 cells, Immunohistochemistry-Paraffin : 1:100-1:500, Immunohistochemistry-Frozen reported by customer review, Proximity Ligation Assay, Immunocytochemistry, CyTOF-ready, Knockout Validated
Formulation PBS
Gene Alias HHCPA78, THIF, thioredoxin binding protein 2, thioredoxin interacting protein, Thioredoxin-binding protein 2, thioredoxin-interacting protein, upregulated by 1,25-dihydroxyvitamin D-3, VDUP1EST01027, Vitamin D3 up-regulated protein 1
Gene Symbols TXNIP
Host Species Mouse
Immunogen Human recombinant TXNIP
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10628
Test Specificity Please note that this antibody is reactive to Mouse and derived from the same host, Mouse. Additional Mouse on Mouse blocking steps may be required for IHC and ICC experiments. Please contact Technical Support for more information.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.