Learn More
TYK2 Mouse anti-Human, Unconjugated, Clone: 1E1A10, Proteintech
Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 674111IG150UL
Description
The TYK2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TYK2. This antibody recognizes human antigen. The TYK2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
TYK2 | |
Monoclonal | |
800 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, ELISA | |
1E1A10 | |
Unconjugated | |
150 μL | |
7297 | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.