Learn More
TYK2 Mouse anti-Human, Unconjugated, Clone: 1E1A10, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | TYK2 |
---|---|
Clone | 1E1A10 |
Concentration | 800 μg/mL |
Applications | Western Blot, Immunohistochemistry, ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-9515
![]() ![]() |
Proteintech Group Inc
674111IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The TYK2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TYK2. This antibody recognizes human antigen. The TYK2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
TYK2 | |
800 μg/mL | |
Monoclonal | |
Mouse | |
7297 | |
Primary |
1E1A10 | |
Western Blot, Immunohistochemistry, ELISA | |
Unconjugated | |
Human | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.