Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tyloxapol, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 1.1000g/mL |
| pH | 4.0 to 7.0 (5% aq. soln.) |
| Flash Point | >113°C |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Quantity | 5 g |
| Specific Gravity | 1.1 |
| Merck Index | 15, 10014 |
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in benzene, toluene, chloroform, carbon, tetrachloride, carbon disulfide, acetic acid |
| Physical Form | Viscous Liquid |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. May cause respiratory irritation. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement IF ON SKIN. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable |
| RTECSNumber | SM9750000 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction