Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tyloxapol, USP, Spectrum™ Chemical
SDP

Catalog No. 18607622 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-622 100 g Amber Glass Bottle
18-607-619 1 g Amber Glass Bottle
18-607-620 5 g Amber Glass Bottle
18-607-621 25 g Amber Glass Bottle
18-607-623 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-607-622 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T1708100GMBL
Only null left
Add to Cart
Add to Cart

T1708, 25301-02-4

Spectrum™ Chemical Tyloxapol, USP

Chemical Identifiers

CAS 25301-02-4
Molecular Formula C17H28O3
Molecular Weight (g/mol) 280.41
InChI Key MDYZKJNTKZIUSK-UHFFFAOYSA-N
IUPAC Name 4-(2,4,4-trimethylpentan-2-yl)phenol; formaldehyde; oxirane
SMILES C=O.C1CO1.CC(C)(C)CC(C)(C)C1=CC=C(O)C=C1

Specifications

CAS Max % 1
pH 4.0 to 7.0
Ignition Residue 0.01
Packaging Amber Glass Bottle
Quantity 100 g
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.