Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TECHREF™ Type RP-1118 Thermal Printer Paper
SDP

Catalog No. 11022104 Shop All TECHREF Inc. Products
Change view
Click to view available options
Type:
Thermal Printer Paper
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
11-022-104 Thermal Printer Paper
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-022-104 Supplier TECHREF™ Supplier No. RP1118
Add to Cart
Add to Cart

Techref Type RP-1118 Thermal Printer Paper is used with HORIBA SLFA single load and turntable XRF analyzers.

  • Superior in contrast and readability with black printing on white background
  • Each roll has a diameter of approx. 48 mm is 80 mm wide and 25 m in length
  • BPA Free

Recommended Storage

Store away from sunlight, in ambient temperatures

Specifications

Type Thermal Printer Paper
Length (Metric) 25 m
Color White
For Use With (Equipment) HORIBA SLFA Single Load and Turntable XRF Analyzers
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.