Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tyrosinase Antibody (TYR/2024R), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB383798 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB383798 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB383798 Supplier Novus Biologicals Supplier No. NBP308892AF594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Tyrosinase Monoclonal antibody specifically detects Tyrosinase in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Tyrosinase
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone TYR/2024R
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias CMM8, EC 1.14.18.1, LB24-AB, Monophenol monooxygenase, OCA1A, OCAIA, SHEP3, SK29-AB, Tumor rejection antigen AB, tyrosinase, tyrosinase (oculocutaneous albinism IA)
Host Species Rabbit
Immunogen Recombinant human full-length tyrosinase (TYR) protein (Uniprot: P14679)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 7299
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.