Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tyrosine Hydroxylase Rabbit anti-Drosophila, Human, Insect, Mammalia, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB30010925 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB30010925 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB30010925 Supplier Novus Biologicals Supplier No. NB30010925UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 147 publications

Tyrosine Hydroxylase Polyclonal specifically detects Tyrosine Hydroxylase in Human, Mouse, Rat, Drosophila, Insect, Mammal samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating, Immunohistochemistry Whole-Mount, Knockout Validated, Dual RNAscope ISH-IHC.

Specifications

Antigen Tyrosine Hydroxylase
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Simple Western 2 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:1000, Immunohistochemistry Free-Floating, Immunocytochemistry 1:500
Formulation 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative
Gene Accession No. P07101
Gene Alias anti-TH, DYT14, DYT5b, EC 1.14.16, EC 1.14.16.2, TH, TH mouse, TYH dystonia 14, Tyrosine 3-hydroxylase, tyrosine 3-monooxygenase, tyrosine hydroxylase, Tyrosine Hydroxylase immunohistochemistry, Tyrosine Hydroxylase mouse
Gene Symbols TH
Host Species Rabbit
Immunogen SDS-denatured, native rat tyrosine hydroxylase purified from pheochromocytoma
Molecular Weight of Antigen 60 kDa
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Neurotransmission, Prostate Cancer, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 7054
Test Specificity Specific for the ∼60k tyrosine hydroxylase protein.
Target Species Human, Mouse, Rat, Drosophila, Insect, Mammalia
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.