Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tyrosylprotein Sulfotransferase 2/TPST2 Antibody, Novus Biologicals™
SDP

Catalog No. NB095661 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB095661 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB095661 Supplier Novus Biologicals Supplier No. NBP303879
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Tyrosylprotein Sulfotransferase 2/TPST2 Polyclonal specifically detects Tyrosylprotein Sulfotransferase 2/TPST2 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Tyrosylprotein Sulfotransferase 2/TPST2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias EC 2.8.2.20, TPST-2, tyrosylprotein phosphotransferase 2, tyrosylprotein sulfotransferase 2protein-tyrosine sulfotransferase 2, tyrosylprotein sulfotransferase-2
Host Species Rabbit
Immunogen Recombinant protein of human TPST2.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8459
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.