Learn More
Description
Specifications
Specifications
| Antigen | TYRP1 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | SPM456 |
| Conjugate | DyLight 650 |
| Dilution | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen |
| Gene Alias | b-PROTEIN, CAS25,6-dihydroxyindole-2-carboxylic acid oxidase, Catalase B, CATB, EC 1.14.18, EC 1.14.18.-, EC 1.14.18.1, Glycoprotein 75, GP75DHICA oxidase, Melanoma antigen gp75, TRP-1, TRPTYRP, tyrosinase-related protein 1TRP1, TYRRP |
| Gene Symbols | TYRP1 |
| Host Species | Mouse |
| Immunogen | SK-MEL-23 cells |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
