Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TYRP1 Antibody (TYRP1/1986), Novus Biologicals™
SDP

Catalog No. NB261599200 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
200 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB261599200 200 μg
NBP26159920 20 μg
NB261599100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB261599200 Supplier Novus Biologicals Supplier No. NBP261599200UG

Mouse Monoclonal Antibody

TYRP1 Monoclonal antibody specifically detects TYRP1 in Human samples. It is validated for Immunohistochemistry-Paraffin, SDS-Page.

Specifications

Antigen TYRP1
Applications Immunohistochemistry (Paraffin), SDS-Page
Classification Monoclonal
Clone TYRP1/1986
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml, SDS-Page
Formulation Prepared in 10mM PBS with 0.05% BSA with 0.5% Sodium Azide
Gene Alias b-PROTEIN, CAS25,6-dihydroxyindole-2-carboxylic acid oxidase, Catalase B, CATB, EC 1.14.18, EC 1.14.18.-, EC 1.14.18.1, Glycoprotein 75, GP75DHICA oxidase, Melanoma antigen gp75, TRP-1, TRPTYRP, tyrosinase-related protein 1TRP1, TYRRP
Gene Symbols TYRP1
Host Species Mouse
Immunogen Recombinant fragment of human TYRP1 protein (around aa 257-377) (exact sequence is proprietary)
Molecular Weight of Antigen 75 kDa
Purification Method Protein A or G purified
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) 7306
Target Species Human
Content And Storage Storage is content dependent.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.