Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TYRP1 Antibody (TYRP1/1986), Novus Biologicals™

Mouse Monoclonal Antibody
$268.00 - $524.00
Specifications
Antigen | TYRP1 |
---|---|
Clone | TYRP1/1986 |
Concentration | 0.2 mg/mL |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml, SDS-Page |
Applications | Immunohistochemistry (Paraffin), SDS-Page |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP26159920
![]() |
Novus Biologicals
NBP26159920UG |
20 μg |
Each for $268.00
|
|
|||||
NB261599100
![]() |
Novus Biologicals
NBP261599100UG |
100 μg |
Each for $524.00
|
|
|||||
NB261599200
![]() |
Novus Biologicals
NBP261599200UG |
200 μg | N/A | N/A | N/A | ||||
Description
TYRP1 Monoclonal specifically detects TYRP1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
TYRP1 | |
0.2 mg/mL | |
Immunohistochemistry (Paraffin), SDS-Page | |
Unconjugated | |
Mouse | |
Prepared in 10mM PBS with 0.05% BSA with 0.5% Sodium Azide | |
7306 | |
Recombinant fragment of human TYRP1 protein (around aa 257-377) (exact sequence is proprietary) | |
Protein A or G purified | |
75 kDa |
TYRP1/1986 | |
Immunohistochemistry-Paraffin 1-2 ug/ml, SDS-Page | |
Monoclonal | |
Purified | |
Human | |
b-PROTEIN, CAS25,6-dihydroxyindole-2-carboxylic acid oxidase, Catalase B, CATB, EC 1.14.18, EC 1.14.18.-, EC 1.14.18.1, Glycoprotein 75, GP75DHICA oxidase, Melanoma antigen gp75, TRP-1, TRPTYRP, tyrosinase-related protein 1TRP1, TYRRP | |
TYRP1 | |
Primary | |
Storage is content dependent. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title