Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TYRP1 Antibody (TYRP1/3281), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB389643 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB389643 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB389643 Supplier Novus Biologicals Supplier No. NBP308897AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TYRP1 Monoclonal antibody specifically detects TYRP1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen TYRP1
Applications ELISA, Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone TYRP1/3281
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias b-PROTEIN, CAS25,6-dihydroxyindole-2-carboxylic acid oxidase, Catalase B, CATB, EC 1.14.18, EC 1.14.18.-, EC 1.14.18.1, Glycoprotein 75, GP75DHICA oxidase, Melanoma antigen gp75, TRP-1, TRPTYRP, tyrosinase-related protein 1TRP1, TYRRP
Host Species Mouse
Immunogen A recombinant fragment (around aa 257-377) of human TYRP1 protein (exact sequence is proprietary) (Uniprot: P17643)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 7306
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.