Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
New Pig Tyvek™ IsoClean™ Cleanroom Lab Coat
Shop All New Pig Corporation Products

Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- The lab coat is made from Tyvek™ material, which is a flashspun high-density polyethylene.
- This provides an excellent barrier against dry particulates, microorganisms, and non-hazardous liquid splashes.
- The unique manufacturing process for the Tyvek™ IsoClean™ fabric is designed to minimize both linting and particle shedding, which is a critical requirement for maintaining the integrity of ISO Class 5-8 cleanroom environments.
- The Tyvek™ material is known for its high durability and resistance to tearing and abrasion, ensuring the lab coat remains intact and protective during use.
- The nonwoven structure of Tyvek™ allows air and moisture vapor to pass through, which helps to reduce heat stress and keeps the wearer more comfortable during extended periods of use.
- These lab coats are available in both clean-processed and sterile options. The sterile versions are gamma-irradiated to a Sterility Assurance Level (SAL) of -6, making them suitable for sterile compounding and other controlled applications.
- The lab coat typically features a front zipper or snap closure, a collar, and elastic wrists to provide a secure fit and a professional appearance while preventing contamination.
Specifications
Specifications
Product Type | Lab Coat |
Size | Small |
Gender | Unisex |
Material | Tyvek |
Color | White |
Closure Type | Zipper |
Cuff Style | Knit |
Disposable | Single-use |
Sterility | Non-sterile |
Resistance | Microorganisms, Dry Particulates, Splash |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction