Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TYW3 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB357869 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB357869 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB357869 Supplier Novus Biologicals Supplier No. NBP176584MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TYW3 Polyclonal antibody specifically detects TYW3 in Human, Mouse samples. It is validated for Western Blot, ELISA

Specifications

Antigen TYW3
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias C1orf171, chromosome 1 open reading frame 171, EC 2.1.1, EC 2.1.1.-, FLJ40918, tRNA wybutosine-synthesizing protein 3 homolog, tRNA-yW synthesizing protein 3 homolog (S. cerevisiae), tRNA-yW-synthesizing protein 3
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the center of human TYW3. The immunogen is located within amino acids 160 - 210 of TYW3. Amino Acid Squence: ANQKMEENKKRIERFYN
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127253
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.