Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TZFP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113557
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIPA5113557 100 μg
1 options

Catalog No. PIPA5113557

Supplier: Invitrogen™ PA5113557

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

TZFP Polyclonal Antibody for ELISA, IHC (P), Western Blot

DNA-binding protein that binds to the to a 5'-TGTACAGTGT-3' core sequence. May function as a transcriptional transactivator and transcriptional repressor. Probably exerts its repressor effect by preventing GATA3 from binding to DNA. May play a role in regulating the differentiation and activation of helper T-cells. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen TZFP
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ZBTB32
Gene Accession No. Q9JKD9, Q9Y2Y4
Gene Alias 4930524C15Rik; FANCC-interacting protein; Fanconi anemia zinc finger protein; FAXF; FAZF; GATA repressor; OTTHUMP00000045790; PLZP; repressor of GATA; Rog; Testis zinc finger protein; TZFP; ZBTB32; zinc finger and BTB domain containing 32; zinc finger and BTB domain-containing protein 32; Zinc finger protein 538; ZNF538
Gene Symbols ZBTB32
Host Species Rabbit
Immunogen Recombinant Human Zinc finger and BTB domain-containing protein 32 protein (1-294AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27033, 58206
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.