Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

u-Plasminogen Activator/Urokinase Antibody (OTI5H4), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. N274793A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274793A647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274793A647 Supplier Novus Biologicals Supplier No. NBP274793AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

u-Plasminogen Activator/Urokinase Monoclonal specifically detects u-Plasminogen Activator/Urokinase in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen u-Plasminogen Activator/Urokinase
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5H4
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ATF, EC 3.4.21, EC 3.4.21.73, plasminogen activator, urokinase, u-PA, UPA, U-plasminogen activator, urinary, urokinase-type plasminogen activator
Gene Symbols PLAU
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 107-379 of human u-Plasminogen Activator/Urokinase (NP_002649) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5328.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.