Learn More
u-Plasminogen Activator/Urokinase Antibody (U-16) - BSA Free, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | u-Plasminogen Activator/Urokinase |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry, ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | U-16 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
| Formulation | 10mM Phosphate (pH 7.4) and 0.5M NaCl with 0.05% Sodium Azide |
| Gene Alias | ATF, EC 3.4.21, EC 3.4.21.73, plasminogen activator, urokinase, uPA, u-PA, U-plasminogen activator, urinary, urokinase-type plasminogen activator |
| Gene Symbols | PLAU |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.