Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
u-Plasminogen Activator/Urokinase Antibody (U-16) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50
Specifications
Antigen | u-Plasminogen Activator/Urokinase |
---|---|
Clone | U-16 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Immunohistochemistry (Frozen), Immunohistochemistry, ELISA, Western Blot |
Description
u-Plasminogen Activator/Urokinase Monoclonal specifically detects u-Plasminogen Activator/Urokinase in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
u-Plasminogen Activator/Urokinase | |
1.0 mg/mL | |
Immunohistochemistry (Frozen), Immunohistochemistry, ELISA, Western Blot | |
Unconjugated | |
Mouse | |
Cancer | |
5328 | |
Native human uPA | |
Primary | |
Store at 4C in the dark. |
U-16 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
PLAU | |
IgG1 | |
Protein A or G purified | |
It is specific for human uPA. No reaction with human tissue plasminogen activator (tPA) when tested by ELISA, immunoblotting and enzyme inhibition and no reaction with any other human plasma proteins when tested by immunoblotting. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title