Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

U0126, 99+%, PROMO, Thermo Scientific Chemicals

Catalog No. J61246MBPM
Click to view available options
Quantity:
25 mg
100 mg

Selective inhibitor of mitogen-activated protein kinases MEK-1 and MEK-2

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109511-58-2
Molecular Formula C18H16N6S2
Synonym 1,4-Diamino-2,3-dicyano-1,4-bis(2-aminophenylthio)butadiene

Specifications

Quantity 25 mg
Content And Storage -30°C to -10°C
For Use With (Application) Selective inhibitor of mitogen-activated protein kinases MEK-1 and MEK-2
Solubility Soluble in DMSO; Poorly soluble in ethanol and water
Product Type Signal Transduction Reagent
Form Powder
Assay ≥99%

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.