Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

U1A Antibody, Novus Biologicals™
SDP

Catalog No. NBP253095 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP253095 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP253095 Supplier Novus Biologicals Supplier No. NBP253095
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

U1A Polyclonal specifically detects U1A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen U1A
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:3000, Immunocytochemistry/Immunofluorescence 1:100 - 1:1000, Immunohistochemistry-Paraffin 1:100 - 1:1000
Formulation PBS (pH 7.0), 1.0% BSA, and 20% Glycerol with 0.01% Thimerosal
Gene Alias Mud1, small nuclear ribonucleoprotein polypeptide A, U1 snRNP A, U1 snRNP-specific protein A, U1-AU1 small nuclear ribonucleoprotein A, U1AU1 small nuclear RNP-specific A
Gene Symbols SNRPA
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human SNRPA. The exact sequence is proprietary.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6626
Target Species Human, Mouse, Rat, Pig, Bovine, Monkey, Rabbit, Sheep, Zebrafish
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.