Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

U2AF2 Antibody, Novus Biologicals™
SDP

Catalog No. NB1065676PC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1065676PC 20 μL
NBP157251 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB1065676PC Supplier Novus Biologicals Supplier No. NBP15725120UL

Rabbit Polyclonal Antibody

U2AF2 Polyclonal specifically detects U2AF2 in Human samples. It is validated for Western Blot.

Specifications

Antigen U2AF2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q96HC5
Gene Alias hU2AF(65), hU2AF65, splicing factor U2AF 65 kD subunit, splicing factor U2AF 65 kDa subunit, U2 (RNU2) small nuclear RNA auxiliary factor 2, U2 auxiliary factor 65 kDa subunit, U2 small nuclear ribonucleoprotein auxiliary factor (65kD), U2 small nuclear RNA auxiliary factor 2, U2AF65U2 snRNP auxiliary factor large subunit
Gene Symbols U2AF2
Host Species Rabbit
Immunogen Synthetic peptides corresponding to U2AF2 (U2 small nuclear RNA auxiliary factor 2) The peptide sequence was selected from the N terminal of U2AF2. Peptide sequence EFERQLNENKQERDKENRHRKRSHSRSRSRDRKRRSRSRDRRNRDQRSAS.
Molecular Weight of Antigen 52 kDa
Purification Method Protein A purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11338
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.