Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
U2AF2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
| Antigen | U2AF2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB1065676PC
![]() |
Novus Biologicals
NBP15725120UL |
20 μL |
Each for $206.00
|
|
|||||
NBP157251
![]() |
Novus Biologicals
NBP157251 |
100 μL |
Each for $487.50
|
|
|||||
Description
U2AF2 Polyclonal specifically detects U2AF2 in Human samples. It is validated for Western Blot.Specifications
| U2AF2 | |
| Polyclonal | |
| Purified | |
| RUO | |
| hU2AF(65), hU2AF65, splicing factor U2AF 65 kD subunit, splicing factor U2AF 65 kDa subunit, U2 (RNU2) small nuclear RNA auxiliary factor 2, U2 auxiliary factor 65 kDa subunit, U2 small nuclear ribonucleoprotein auxiliary factor (65kD), U2 small nuclear RNA auxiliary factor 2, U2AF65U2 snRNP auxiliary factor large subunit | |
| U2AF2 | |
| IgG | |
| Protein A purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Q96HC5 | |
| 11338 | |
| Synthetic peptides corresponding to U2AF2 (U2 small nuclear RNA auxiliary factor 2) The peptide sequence was selected from the N terminal of U2AF2. Peptide sequence EFERQLNENKQERDKENRHRKRSHSRSRSRDRKRRSRSRDRRNRDQRSAS. | |
| Primary | |
| 52 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title