Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

U2AF2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220775 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220775 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220775 Supplier Novus Biologicals Supplier No. NBP220775
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

U2AF2 Polyclonal specifically detects U2AF2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen U2AF2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.79mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000-1:20000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. P26368
Gene Alias hU2AF(65), hU2AF65, splicing factor U2AF 65 kD subunit, splicing factor U2AF 65 kDa subunit, U2 (RNU2) small nuclear RNA auxiliary factor 2, U2 auxiliary factor 65 kDa subunit, U2 small nuclear ribonucleoprotein auxiliary factor (65kD), U2 small nuclear RNA auxiliary factor 2, U2AF65U2 snRNP auxiliary factor large subunit
Gene Symbols U2AF2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human U2AF65. The exact sequence is proprietary.
Molecular Weight of Antigen 53 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11338
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.