Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

U2AF2 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB290567 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290567 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290567 Supplier Novus Biologicals Supplier No. NBP204139PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

U2AF2 Polyclonal antibody specifically detects U2AF2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation

Specifications

Antigen U2AF2
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation
Formulation PBS
Gene Alias hU2AF(65), hU2AF65, splicing factor U2AF 65 kD subunit, splicing factor U2AF 65 kDa subunit, U2 (RNU2) small nuclear RNA auxiliary factor 2, U2 auxiliary factor 65 kDa subunit, U2 small nuclear ribonucleoprotein auxiliary factor (65kD), U2 small nuclear RNA auxiliary factor 2, U2AF65U2 snRNP auxiliary factor large subunit
Host Species Rabbit
Immunogen Maps to a region between residue 380 and 430 of human U2 Small Nuclear Ribonucleoprotein Auxiliary Factor (65kD) using the numbering given in entry NP_009210.1 (GeneID 11338).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11338
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.